Learn More
ESRP1, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA554873
Description
RBM35A, also known as ESRP1, is a mRNA splicing factor that with its related protein RBM35B (ESRP2) are coordinators of an epithelial cell-type-specific splicing program. RBM35A contains three putative RNA recognition motifs and acts by directly binding specific sequences in mRNAs. RBM35A is involved in posttranscriptional regulation of a number of genes such as FGFR2, CD44, CTNND1, and ENAH by exerting a differential effect on protein translation via 5' UTRs of mRNAs. Other recent studies have shown that RMB35A may also act as a novel tumor suppressor.
Immunogen sequence: QLHVRQILHP EASKKNVLLP ECFYSFFDLR KEFKKCCPGS PDIDKLDVAT MTEYLNFEKS SSVSRYGASQ VEDMGNIILA MISEPYNHRF SDPERVNYKF ESHighest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 93%.
Specifications
| ESRP1 | |
| Polyclonal | |
| Unconjugated | |
| ESRP1 | |
| RBM35A, RMB35A, RNA binding motif protein 35A, RNA-binding motif protein 35A, RNA-binding protein 35A | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 54845 | |
| Store at 4°C short term. Store at −20°C long term. Avoid freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry | |
| 0.1mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q6NXG1 | |
| ESRP1 | |
| Recombinant protein corresponding to Human ESRP1 | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Research Use Only