Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ESRP2 Antibody (22C5.H7.A6), Alexa Fluor™ 700, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP177972AF700
Description
ESRP2 Monoclonal specifically detects ESRP2 in Mouse samples. It is validated for Western Blot, ELISA.Specifications
ESRP2 | |
Monoclonal | |
Alexa Fluor 700 | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
80004 | |
Store at 4°C in the dark. | |
IgG2a κ |
Western Blot, ELISA | |
22C5.H7.A6 | |
Western Blot, ELISA | |
epithelial splicing regulatory protein 2, FLJ21918, FLJ22248, RBM35BRNA-binding protein 35B, RNA binding motif protein 35A, RNA binding motif protein 35B, RNA-binding motif protein 35B | |
ESRP2 Antibody (22C5.H7.A6) is produced by repeated immunizations of full length recombinant mouse Esrp-2 fusion protein. (Uniprot: Q8K0G8) | |
0.1 mL | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction