Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ESRP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583919
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583919 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583919 Supplier Invitrogen™ Supplier No. PA583919
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TTVGYLTTPT AALASAPTSV LSQSGALVRM QGVPYTAGMK DLLSVFQAYQ LPADDYTSLM PVGDPPRTVL QAPKEWVCL Highest antigen sequence indentity to the following orthologs: Rat - 87%, Mouse - 89%.

RBM35A, also known as ESRP2, is a mRNA splicing factor that with its related protein RBM35A (ESRP1) are coordinators of an epithelial cell-type-specific splicing program. Both RBM35B and RBM35A are involved in posttranscriptional regulation of a number of genes such as FGFR2, CD44, CTNND1, and ENAH by exerting a differential effect on protein translation via 5' UTRs of mRNAs, suggesting that these proteins are global regulators of an epithelial regulatory network. Loss of this global ESRP-regulated epithelial splicing program induces the phenotypic changes in cell morphology that are observed during the epithelial-mesenchymal transition.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ESRP2
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ESRP2
Gene Accession No. Q9H6T0
Gene Alias 9530027K23Rik; Epithelial splicing regulatory protein 2; ESRP2; PP7059; RBM35B; RGD1310855; RNA binding motif protein 35A; RNA binding motif protein 35B; RNA-binding motif protein 35B; RNA-binding protein 35B
Gene Symbols ESRP2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ESRP2. Recombinant protein control fragment (Product #RP-104859).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80004
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.