Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ESRP2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA597717
Description
ESRP2 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA
RBM35A, also known as ESRP2, is a mRNA splicing factor that with its related protein RBM35A (ESRP1) are coordinators of an epithelial cell-type-specific splicing program. Both RBM35B and RBM35A are involved in posttranscriptional regulation of a number of genes such as FGFR2, CD44, CTNND1, and ENAH by exerting a differential effect on protein translation via 5' UTRs of mRNAs, suggesting that these proteins are global regulators of an epithelial regulatory network. Loss of this global ESRP-regulated epithelial splicing program induces the phenotypic changes in cell morphology that are observed during the epithelial-mesenchymal transition.
Specifications
ESRP2 | |
Polyclonal | |
Unconjugated | |
ESRP2 | |
9530027K23Rik; Epithelial splicing regulatory protein 2; ESRP2; PP7059; RBM35B; RGD1310855; RNA binding motif protein 35A; RNA binding motif protein 35B; RNA-binding motif protein 35B; RNA-binding protein 35B | |
Rabbit | |
Protein G | |
RUO | |
80004 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
3.06 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
Q9H6T0 | |
ESRP2 | |
Recombinant Human Epithelial splicing regulatory protein 2 protein (615-720AA). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction