Learn More
ESRRA Mouse anti-Human, Clone: OTI3G10, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF809108

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Estrogen-related receptor alpha (ERR alpha), a NR3 Steroid Receptor, was isolated based on sequence similarity in its DNA-binding domain to estrogen receptor alpha (ER alpha). ERR alpha has been shown to regulate the promoters of lactoferrin, medium-chain acyl CoA dehydrogenase, osteopontin, and thyroid receptor alpha, and it may affect cellular energy balance and bone formation.Specifications
ESRRA | |
Monoclonal | |
Unconjugated | |
ESRRA | |
ERR1, ERRa, ERRalpha, ESRL1, NR3B1 | |
Mouse | |
Affinity Chromatography | |
RUO | |
2101 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI3G10 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P11474 | |
ESRRA | |
Human recombit protein fragment corresponding to amino acids 206-417 of human ESRRA produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.