Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ESRRA Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA533521
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA533521 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA533521 Supplier Invitrogen™ Supplier No. PA533521
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Monkey, Marmoset, Mouse, Rat, Hamster, Elephant, Panda, Bovine, Bat, Dog, Horse, Pig (100%).

Estrogen-related receptor alpha 1 (ERR alpha, ERR1) is an orphan member of the nuclear hormone receptor superfamily. All the ERR family members share an almost identical DNA-binding domain, which has 68% amino acid identity with that of estrogen receptor. This protein binds to an ERR-alpha response element (ERRE) containing a single consensus half-site, 5'-tnaaggtca-3' can bind to the medium-chain acyl coenzyme a dehydrogenase (mcad) response element nrre-1 and may act as an important regulator of mcad promoter. Studies have shown that ERR1 functions a metabolic regulator and is often expressed in tissues that preferentially metabolize fatty acids. The corresponding gene for the ERR1 is NR3B1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ESRRA
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene ESRRA
Gene Accession No. O08580, P11474, Q5QJV7, Q6QMY5
Gene Alias ERR alpha; ERR1; ERRa; ERRalpha; ERR-alpha; Errra; ESRL1; ESRRA; ESRRA1; ESRRA2; estrogen receptor related 1; estrogen receptor-like 1; estrogen related receptor alpha; estrogen related receptor alpha 1; estrogen related receptor, alpha; estrogen-related nuclear receptor alpha; estrogen-related receptor alpha; estrogen-related receptor, alpha; Estrra; HERR1; LOW QUALITY PROTEIN: steroid hormone receptor ERR1; NR3B1; Nuclear receptor subfamily 3 group B member 1; orphan nuclear receptor; steroid hormone receptor ERR1; Steroid hormone receptor ERR1 (Estrogen-related receptor, alpha) (ERR-alpha) (Estrogen receptor-like 1)
Gene Symbols ESRRA
Host Species Rabbit
Immunogen Synthetic 15 amino acid peptide from ligand-binding domain of human ESRRA.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100055615, 100322868, 100761304, 2101, 26379, 293701, 403169, 507834
Target Species Human, Mouse, Rat, Canine, Monkey, Bovine, Hamster, Pig, Horse, Bat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.