Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ESRRB Monoclonal Antibody (OTI1E6), TrueMAB™, OriGene
SDP

Catalog No. 501680709
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-168-0709 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-168-0709 Supplier OriGene Supplier No. TA805455
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ESRRB (steroid hormone receptor ERR2) plays a role in maintaice of self-renewal and pluripotency of embryonic and trophoblast stem cells through different signaling pathways including FGF signaling pathway and Wnt signaling pathways. Upon FGF signaling pathway activation ESRRB interacts with KDM1A by directly binding to enhancer sites of ELF5 and EOMES and activates their transcription leading to self-renewal of trophoblast stem cells. ESSRB also regulates expression of multiple rod-specific genes and is required for survial of this cell type. Defects in the gene can cause deafness, autosomal recessive, 35 (DFNB35).

Specifications

Antigen ESRRB
Applications Western Blot
Classification Monoclonal
Clone OTI1E6
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene ESRRB
Gene Accession No. O95718
Gene Alias DFNB35, ERR2, ERRb, ESRL2, NR3B2
Gene Symbols ESRRB
Host Species Mouse
Immunogen Human recombit protein fragment corresponding to amino acids 1-252 of human ESRRB produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2103
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.