Learn More
Invitrogen™ ESRRB Polyclonal Antibody

Description
ESRRB Polyclonal Antibody for Western Blot, IHC (P), Flow
Specifications
Specifications
| Antigen | ESRRB |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | ESRRB |
| Gene Accession No. | O95718 |
| Gene Alias | DFNB35; ERR beta-2; ERR2; Err-2; ERRb; ERRB2; ERRbeta; ERR-beta; ERRbeta-2; ESRL2; ESRRB; estrogen receptor related 2; estrogen receptor-like 2; estrogen related receptor beta; estrogen related receptor, beta; estrogen-related nuclear receptor beta; estrogen-related receptor beta; estrogen-related receptor, beta; Estrrb; NR3B2; nuclear receptor ERRB2; nuclear receptor subfamily 3 group B member 2; orphan nuclear receptor; steroid hormone receptor ERR2 |
| Gene Symbols | ESRRB |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.