Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ESRRB Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA533522
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA533522 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA533522 Supplier Invitrogen™ Supplier No. PA533522
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Percent identity with other species by BLAST analysis: Human, Gorilla, Monkey, Marmoset, Hamster, Panda, Horse (100%) Mouse, Rat, Elephant, Bovine, Bat, Rabbit, Pig (93%) Dog (86%).

ESRRB (steroid hormone receptor ERR2) plays a role in maintenance of self-renewal and pluripotency of embryonic and trophoblast stem cells through different signaling pathways including FGF signaling pathway and Wnt signaling pathways. Upon FGF signaling pathway activation ESRRB interacts with KDM1A by directly binding to enhancer sites of ELF5 and EOMES and activates their transcription leading to self-renewal of trophoblast stem cells. ESSRB also regulates expression of multiple rod-specific genes and is required for survival of this cell type. Defects in the gene can cause deafness, autosomal recessive, 35 (DFNB35).
TRUSTED_SUSTAINABILITY

Specifications

Antigen ESRRB
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene ESRRB
Gene Accession No. O95718
Gene Alias DFNB35; ERR beta-2; ERR2; Err-2; ERRb; ERRB2; ERRbeta; ERR-beta; ERRbeta-2; ESRL2; ESRRB; estrogen receptor related 2; estrogen receptor-like 2; estrogen related receptor beta; estrogen related receptor, beta; estrogen-related nuclear receptor beta; estrogen-related receptor beta; estrogen-related receptor, beta; Estrrb; NR3B2; nuclear receptor ERRB2; nuclear receptor subfamily 3 group B member 2; orphan nuclear receptor; steroid hormone receptor ERR2
Gene Symbols ESRRB
Host Species Rabbit
Immunogen Synthetic 14 amino acid peptide from internal region of human ESRRB.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100058699, 100760486, 2103
Target Species Human, Monkey, Hamster, Horse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.