Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ESRRG Monoclonal Antibody (OTI1D10), TrueMAB™, OriGene
SDP

Catalog No. 502540004
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-0004 30 μL
50-167-5539 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-0004 Supplier OriGene Supplier No. TA504963S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ESRRG Monoclonal antibody specifically detects ESRRG in Human samples. It is validated for Immunocytochemistry, Western Blot

Specifications

Antigen ESRRG
Applications Immunocytochemistry, Western Blot
Classification Monoclonal
Clone OTI1D10
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene Rab3il1
Gene Accession No. Q8TBN0
Gene Alias DKFZp781L1617; ERR gamma; ERR gamma-2; ERR3; errg; ERRG2; ERRgamma; ESRRG; Estrogen receptor-related protein 3; estrogen related receptor gamma; estrogen-related receptor 3; estrogen-related receptor gamma; FLJ16023; Kiaa0832; mKIAA0832; NR3B3; Nuclear receptor subfamily 3 group B member 3
Host Species Mouse
Immunogen Full length human recombinant protein of human RAB3IL1 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5866
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.