Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ESRRG Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA533524
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA533524 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA533524 Supplier Thermo Scientific Supplier No. PA533524
Add to Cart
Add to Cart

ESRRG Polyclonal Antibody

ESRRG Polyclonal Antibody for IHC (P)

TRUSTED_SUSTAINABILITY

Specifications

Antigen ESRRG
Applications ChIP Assay, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene Esrrg
Gene Accession No. P62508, P62509, P62510
Gene Alias DKFZp781L1617; ERR gamma; ERR gamma-2; ERR3; errg; ERRG2; ERRgamma; ESRRG; Estrogen receptor-related protein 3; estrogen related receptor gamma; estrogen-related receptor 3; estrogen-related receptor gamma; FLJ16023; Kiaa0832; mKIAA0832; NR3B3; Nuclear receptor subfamily 3 group B member 3
Gene Symbols Esrrg
Host Species Rabbit
Immunogen Synthetic 15 amino acid peptide from C-terminus of human ESRRG / ERR3.
Purification Method Antigen Affinity Chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100050530, 100355568, 100622056, 100760533, 2104, 26381, 360896, 421357, 478961, 536732
Target Species Human, Mouse, Rat, Canine, Rabbit, Monkey, Bovine, Hamster, Pig, Horse, Chicken
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.