Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ESRRG Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA533525
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA533525 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA533525 Supplier Invitrogen™ Supplier No. PA533525
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ESRRG Polyclonal Antibody for IHC (P)

Estrogen-related receptor gamma (ERRG, ERR3, NR3B3) is an orphan member of the nuclear hormone receptor superfamily. ERRG acts as a transcription activator in the absence of a bound ligand. It binds specifically to an estrogen response element and activates reporter genes controlled by estrogen response elements. ERRG also induces the expression of PERM1 in the skeletal muscle.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ESRRG
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene Esrrg
Gene Accession No. P62508, P62509, P62510
Gene Alias DKFZp781L1617; ERR gamma; ERR gamma-2; ERR3; errg; ERRG2; ERRgamma; ESRRG; Estrogen receptor-related protein 3; estrogen related receptor gamma; estrogen-related receptor 3; estrogen-related receptor gamma; FLJ16023; Kiaa0832; mKIAA0832; NR3B3; Nuclear receptor subfamily 3 group B member 3
Gene Symbols Esrrg
Host Species Rabbit
Immunogen Synthetic 14 amino acid peptide from N-terminus of human ESRRG / ERR3.
Purification Method Antigen Affinity Chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100050530, 100355568, 100760533, 2104, 26381, 360896, 478961, 536732
Target Species Human, Mouse, Rat, Canine, Rabbit, Monkey, Bovine, Hamster, Horse, Bat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.