Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ESRRG Recombinant Rabbit Monoclonal Antibody (4H7L11)

Catalog No. PI702406
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI702406 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI702406 Supplier Invitrogen™ Supplier No. 702406
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

This antibody is predicted to react with Monkey, Sheep, Mouse Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.

Estrogen-related receptor gamma (ERRG, ERR3, NR3B3) is an orphan member of the nuclear hormone receptor superfamily. ERRG acts as a transcription activator in the absence of a bound ligand. It binds specifically to an estrogen response element and activates reporter genes controlled by estrogen response elements. ERRG also induces the expression of PERM1 in the skeletal muscle.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ESRRG
Applications Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 4H7L11
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.2-7.4
Gene Esrrg
Gene Accession No. P62508, P62509, P62510
Gene Alias DKFZp781L1617; ERR gamma; ERR gamma-2; ERR3; errg; ERRG2; ERRgamma; ESRRG; Estrogen receptor-related protein 3; estrogen related receptor gamma; estrogen-related receptor 3; estrogen-related receptor gamma; FLJ16023; Kiaa0832; mKIAA0832; NR3B3; Nuclear receptor subfamily 3 group B member 3
Gene Symbols Esrrg
Host Species Rabbit
Immunogen Peptides corresponding to human ESRRG (aa245-263, 318-333).
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2104, 26381, 360896
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.