Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cole-Parmer™ Essentials Disposable Plastic Lab Forceps

Catalog No. 03400506 Shop All Oakton Products
Change view
Click to view available options
Length (Metric):
12.2 cm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
03-400-506 12.2 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 03-400-506 Supplier Cole-Parmer™ Supplier No. 0644320
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The corrugated handles of these plastic forceps provide a secure finger grip for better control.

  • Being disposable reduces cross contamination
  • Serrated tips for accurate sample grip
  • Sterile and individually bagged
  • All plastic forceps have corrugated handles to provide you with a secure finger grip
  • Use serrated pointed tips for handling smaller objects or serrated blunt tips for general use

Specifications

Length (English) 4.8 in.
Length (Metric) 12.2 cm
Material Polystyrene
Quantity 100 Forceps/Cs.
Sterility Sterile
Style Micro Alligator Ear
Tip Style Serrated
Product Type Forceps
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.