Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Estradiol Antibody (AB5.1), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP178621
Description
Estradiol Monoclonal specifically detects Estradiol in Mouse, Non-species specific samples. It is validated for ELISA, Immunoprecipitation, Radioimmunoassay.Specifications
| Estradiol | |
| Monoclonal | |
| 1 mg/mL | |
| ELISA 1:100 - 1:2000, Immunoprecipitation 1:10 - 1:500, Radioimmunoassay 1:100 - 1:2000 | |
| Estradiol-6-CMO-KLH | |
| 100 μg | |
| Primary | |
| Other | |
| IgG1 |
| ELISA, Immunoprecipitation, Radioimmune Assays (RIA) | |
| AB5.1 | |
| Unconjugated | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Chemical. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction