Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

β-Estradiol, MP Biomedicals

Catalog No. mp021016562 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP021016562 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP021016562 Supplier MP Biomedicals, Inc Supplier No. 02101656.2
Only null left
Add to Cart
Add to Cart

  • β-Estradiol is used to study cell differentiation and transformations (tumorigenicity)
  • The pharmacology and HPLC determination of β-Estradiol in plasma and urine have been reported

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50-28-2
Molecular Formula C18H24O2
Molecular Weight (g/mol) 272.388
MDL Number MFCD00003693
InChI Key VOXZDWNPVJITMN-ZBRFXRBCSA-N
Synonym estradiol, beta-estradiol, 17beta-estradiol, oestradiol, dihydrofolliculin, estrace, ovocyclin, progynon, dihydrotheelin, dihydroxyestrin
PubChem CID 5757
ChEBI CHEBI:16469
IUPAC Name (8R,9S,13S,14S,17S)-13-methyl-6,7,8,9,11,12,14,15,16,17-decahydrocyclopenta[a]phenanthrene-3,17-diol
SMILES CC12CCC3C(C1CCC2O)CCC4=C3C=CC(=C4)O

Specifications

Melting Point 173°C to 179°C
Color White
Linear Formula C18H24O2
Quantity 250 mg
Solubility Information Almost insoluble in water(50mg/mL-clear, colorless solution). Freely soluble in alcohol; soluble in acetone, dioxane, other organic solvents; solutions of fixed alkali hydroxides; sparingly soluble in vegetable oils; soluble in methanol:CHCl3 (1:1)
Formula Weight 272.39
Physical Form Powder
Chemical Name or Material β-ESTRADIOL
Recommended Storage Store at Room Temperature (15-30°C). Store Under Nitrogen.
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.