Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ α-Estradiol
GREENER_CHOICESDP

Catalog No. 282350 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
28-235-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 28-235-0 Supplier Tocris Bioscience™ Supplier No. 2823/50
Add to Cart
Add to Cart
This item is not returnable. View return policy

Endogenous estrogen receptor agonist

Endogenous estrogen receptor ligand (Ki values are 0.2 and 1.2 nM for ERα and ERβ receptors respectively).

β-Estradiol also available.

Chemical Identifiers

CAS 57-91-0
Molecular Formula C18H24O2

Specifications

Quantity 50 mg
Formula Weight 272.38
Purity >96%
Molecular Weight (g/mol) 272.388
Chemical Name or Material α-Estradiol
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.