Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Estriol ELISA Kit

Catalog No. p-200006749
Encompass_Preferred
Click to view available options
Quantity:
10 x 96 Tests
96 Tests

ELISA

The Estriol Competitive ELISA quantitates estriol in serum, plasma, urine, saliva, dried fecal extracts or or cell culture medium.∣Principle of the method: The Estriol Competitive ELISA research-use-only kit is designed to quantitatively measure estriol independent of species. An estriol standard is provided to generate a standard curve for the assay and all samples are read off a user-generated standard curve. Standards or diluted samples are pipetted into a clear microtiter plate coated with anti-rabbit IgG antibodies. Anti-rabbit estriol antibodies and estriol-peroxidase conjugate is added to the wells. The estriol-peroxidase conjugate and any estriol in the sample will compete to bind to the anti-rabbit estriol antibodies. After incubation, the plate is washed and substrate is added. The substrate reacts with the bound estriol-peroxidase conjugate. After a 30 min incubation, the reaction is stopped and the intensity of the generated color is detected in a microtiter plate reader at 450 nm. The intensity of the generated color corresponds inversely to the amount of estriol present in the sample.∣Rigorous validation: Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

Estriol is the main estrogen in the blood and urine of pregnant females. Serum estriol determinations reflect the status of the fetus and are used to evaluate fetal distress and placental function.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number C18H24O3; Estratriol; Estriol; Oestriol; Ovestin; Ovestrion
Assay Range 19.66 to 12,000 pg/mL
Assay Sensitivity 5.33 pg/mL
Conjugate HRP
Product Type ELISA
Sample Type Fecal Extract, Plasma, Serum, Supernatant, Urine
For Use With (Equipment) Colorimetric Microplate Reader
Interassay CV 12%
Intraassay CV 10
Kit Contents Anti-IgG coated 96 well plate, Standard, Assay Buffer Concentrate, Capture Antibody, HRP-conjugated Antigen, Wash Buffer Concentrate, TMB Substrate, Stop Solution, Plate Sealer
Label or Dye HRP
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Fecal Extract 50 μL, Plasma 50 μL, Serum 50 μL, Supernatant 50 μL, Urine 12.5 μL
Storage Requirements -20°C
Target Estriol
Test Time 45 mins
Total Assay Time 2 hrs 30 mins
Show More Show Less
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.