Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Estriol, Micronized, USP, 97-102%, Spectrum™ Chemical
SDP

Catalog No. 18602461 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
500 g
1 kg
Packaging:
Amber Glass Bottle
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-602-461 100 g Amber Glass Bottle
18-602-459 1 g Amber Glass Bottle
18-602-460 5 g Amber Glass Bottle
18-602-463 25 g Amber Glass Bottle
18-602-464 500 g Amber Glass Bottle
18-602-462 1 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18-602-461 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. E1011100GMBL
Add to Cart
Add to Cart

E1011, 50-27-1, C18H24O3

Spectrum™ Chemical Estriol, Micronized, USP is a sex hormone that is used in hormone replacement therapy. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 50-27-1
Molecular Formula C18H24O3
Molecular Weight (g/mol) 288.39
InChI Key PROQIPRRNZUXQM-ZXXIGWHRSA-N
IUPAC Name (1R,2R,3aS,3bR,9bS,11aS)-11a-methyl-1H,2H,3H,3aH,3bH,4H,5H,9bH,10H,11H,11aH-cyclopenta[a]phenanthrene-1,2,7-triol
SMILES C[C@]12CC[C@H]3[C@@H](CCC4=CC(O)=CC=C34)[C@@H]1C[C@@H](O)[C@@H]2O

Specifications

CAS Max % 100%
Melting Point 280°C to 285°C
Boiling Point 370.6°C
Ignition Residue 0.1%
Loss on Drying 0.5%
Packaging Amber Glass Bottle
Quantity 100 g
Specific Rotation +54° to +62°
Percent Purity 97 to 102%
Grade USP
Physical Form Crystalline Powder or Lumps
Chemical Name or Material Estriol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.