Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Estrogen Receptor Ab-11 Mouse Monoclonal Antibody, Epredia™

Catalog No. MS354P1 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
Form:
Concentrated
Concentration:
1mg/mL
200μg/mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form Concentration
MS354P1 500 μL Concentrated 200μg/mL
MS354P1ABX 100 μL Concentrated 1mg/mL
MS354PABX 200 μL Concentrated 1mg/mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. MS354P1 Supplier Epredia Supplier No. MS354P1A
Add to Cart
Add to Cart

Obtain accurate, reproducible results in immunohistochemistry and western blotting experiments with Epredia™ Estrogen Receptor Ab-11, Mouse Monoclonal Antibody.

The ER gene consists of more than 140kb of genomic DNA divided into 8 exons and is translated into a protein with six functionally discrete domains required for transcription activation function, binding to estrogen response element (ERE) constitutive dimerization, binding to heat shock proteins, and ligand recognition. The ER is an important regulator of growth and differentiation in the mammary gland. Presence of ER in breast tumors indicates an increased likelihood of response to anti-estrogen (e.g. tamoxifen) therapy.

Host Species: Mouse

Clone: 1D5

Isotype: IgG1/κ

Species Reactivity: Human, Monkey, Mouse and Rat. Others not known.

Epitope: N-terminal domain

Immunogen: Recombinant human estrogen receptor protein

Molecular Weight: 67 kDa

Positive Control: MCF-7 cells, Breast CA

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting

Specifications

Antigen Estrogen Receptor Ab-11
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 1D5
Concentration 200μg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen Recombinant human estrogen receptor protein
Quantity 500 μL
Regulatory Status ASR, IVD
Research Discipline Steroids Hormones and Growth Factors
Primary or Secondary Primary
Target Species Human, Mouse, Non-human Primate, Rat
Form Concentrated
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.