Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Estrogen Receptor, beta (ER beta) Monoclonal Antibody (ERB), Abbomax

Catalog No. 502204148
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-4148 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-4148 Supplier Abbomax Inc Supplier No. 603142
Add to Cart
Edge
Add to Cart

Mouse Monoclonal Antibody

Positive control: Breast carcinoma Cellular location: Nucleus, Cytoplasm, Cell membrane.

Estrogen Receptor, beta (ER beta) Monoclonal antibody specifically detects Estrogen Receptor, beta (ER beta) in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen Estrogen Receptor, beta (ER beta)
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone ERB
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with proprietary stabilizer and 0.01% sodium azide
Gene ESR2
Gene Accession No. Q92731
Gene Alias ER; ER beta; ER[b]; Erb; Erb2; Erbeta; ER-beta; Esr2; ESRB; ESR-BETA; ESTRB; estrogen receptor 2; estrogen receptor 2 (beta); estrogen receptor 2 (ER beta); estrogen receptor 2 beta; estrogen receptor beta; estrogen receptor beta 1 isoform deletion variant; estrogen receptor beta 4; NR3A2; Nuclear receptor subfamily 3 group A member 2; oestrogen receptor beta; steroid receptor
Host Species Mouse
Immunogen A synthetic peptide corresponding to the c-terminus of human Estrogen Receptor Beta
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2100
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.