Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Estrone-3-Sulfate (E1S) Competitive ELISA Kit

Catalog No. EIA17E3S
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 x 96 Tests
96 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EIA17E3S 96 Tests
EIA17E3SX10 10 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. EIA17E3S Supplier Invitrogen™ Supplier No. EIA17E3S
Only null left
Add to Cart
Add to Cart

ELISA

The Human Estrone-3 Sulfate (E1S) ELISA quantitates human estrone-3 Sulfate in human serum,plasma,urine,dried fecal extracts or cell culture medium.

Principle of the method

The Estrone-3-Sulfate (E1S) Competitive ELISA research-use-only kit uses a specifically generated antibody to measure Estrone-3-Sulfate (E1S) in a variety of matrices,including serum,plasma,urine,and fecal samples. The kit will quantitatively measure E1S present in diluted buffer samples and tissue culture media samples. An E1S standard is provided to generate a standard curve for the assay and all samples should be read off the standard curve. Standards or diluted samples are pipetted into a clear microtiter plate coated with an antibody to capture rabbit antibodies. An E1S-peroxidase conjugate is added to the standards and samples in the wells. The binding reaction is initiated by the addition of a polyclonal antibody to E1S to each well. After a 2-hour incubation the plate is washed and substrate is added. The substrate reacts with the bound E1S-peroxidase conjugate. After a short incubation,the reaction is stopped and the intensity of the generated color is detected in a microtiter plate reader capable of measuring at 450 nm.

Rigorous validation

Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity,specificity,precision,and lot-to-lot consistency. See manual for more information on validation.

Estrone-3-sulfate (1, 3, 5(10)-Estratrien-3-ol-17-one sulfate (E1S) is synthesized in the fetal or cotyledonary portion of the placentome. Estrone sulfate, present in plasma in a higher concentration than either unconjugated estrone or estradiol in nonpregnant women and normal men, appears to originate almost entirely from a conjugation of estrone and converted estradiolin nonglandular tissues. Estrone sulfate is only slowly cleared from plasma, thus its concentration does not fluctuate markedly during the day. Estrone sulfate is quantitatively the most important circulating estrogen. Estrone-3-sulfate (E1S) is identical across all species and so this kit should measure estrone-3-sulfate from all sources.
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 40.96 to 4000 pg/mL
Assay Sensitivity 26.4 pg/mL
Description Estrone-3-Sulfate (E1S) Competitive ELISA Kit
Assay Multiplex assay on the Luminex Platform
Conjugate HRP
Detection Method Absorbance at 450 nm
Product Type ELISA
Protein Family Steroid hormone
Sample Type Fecal Extract, Plasma, Serum, Supernatant, Urine
Sufficient For 96 assays
Target Species Human
For Use With (Application) ELISA, Protein Assays, Aktivity Assays, Protein Biology, Reproductive Biology, Inflammation, Oxidative Stress, Metabolism, Cell Signaling, Epigenetics, Hematology, DNA Damage, Cancer Biology, Immunology, Neuroscience, Neurobiology, Steroid Hormone Detecti
For Use With (Equipment) Colorimetric Microplate Reader
Interassay CV 8.5%
Intraassay CV 4.2
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, HRP-Conjugated Detection Antibody, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 96 Tests
Regulatory Status RUO
Research Discipline Reproductive Biology
Sample Volume Fecal Extract 50 μL, Plasma 1 μL, Serum 1 μL, Supernatant 50 μL, Urine 6.25 μL
Storage Requirements -20°C
Target Estrone-3-Sulfate
Test Time 1 hrs 20 mins
Total Assay Time 2 hrs 30 mins
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.