Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ethylbenzene, 99.8%, pure
Click to view available options
Quantity:
25 mL
500 mL
1 L
2.5 L
25 L
Packaging:
Glass bottle
Metal Drum
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | -95.0°C |
Density | 0.8600g/mL |
Boiling Point | 136.0°C |
Flash Point | 15°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 99.8% |
Packaging | Glass bottle |
Molecular Formula | C8H10 |
Refractive Index | 1.4940 to 1.4970 |
Linear Formula | C6H5C2H5 |
Show More |
Safety and Handling
Hazard Category | Signal Word: DANGER! |
Hazard Statement | GHS H Statement: Harmful if inhaled. Highly flammable liquid and vapor. |
Precautionary Statement | Emergency Overview Flammable liquid and vapor. Possible cancer hazard. May cause cancer based on animal data. Aspiration hazard if swallowed - can enter lungs and cause damage. Harmful by inhalation. Inhalation may cause central nervous system effects. May be harmful if swallowed or absorbed through the skin. May cause skin, eye, and respiratory tract irritation. NFPA Health:2 Flammability:3 Instability:0 |
EINECSNumber | 202-849-4 |
RTECSNumber | DA0700000 |
TSCA | TSCA |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction