Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Estrone Competitive ELISA Kit

Catalog No. eia17e1
Encompass_Preferred
Click to view available options
Quantity:
10 x 96 Tests
96 Tests

ELISA

The Multispecies Estrone ELISA quantitates estrone in fecal extracts,urine,and tissue culture media.

Principle of the method The Estrone Competitive ELISA research-use-only kit is designed to quantitatively measure estrone present in extracted dried fecal samples,urine,and tissue culture media samples. The kit is unique as it measures both non-conjugated estrone,estrone-3-sulfate,and estrone 3-glucuronide in urine and fecal samples with almost equal affinity,allowing for non-invasive testing of this steroid. An estrone standard is provided to generate a standard curve for the assay and all samples should be read off the standard curve. Standards or diluted samples are pipetted into a clear microtiter plate coated with an antibody to capture rabbit antibodies. An estrone-peroxidase conjugate is added to the standards and samples in the wells. The binding reaction is initiated by the addition of a polyclonal antibody to estrone to each well. After a 2-hour incubation the plate is washed and substrate is added. The substrate reacts with the bound estrone-peroxidase conjugate. After a short incubation,the reaction is stopped and the intensity of the generated color is detected in a microtiter plate reader capable of measuring at 450 nm.

Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity,specificity,precision,and lot-to-lot consistency. See manual for more information on validation.

Estrone is an aromatized C18 steroid with a 3-hydroxyl group and a 17-ketone, a major mammalian estrogen. it is converted from androstenedione directly, or from testosterone via estradiol. In humans, it is produced primarily by the cyclic ovaries, placenta, and the adipose tissue of men and postmenopausal women. Therapeutic estrone is the synthetic form of a naturally occurring estrogen estrone. Estrone diffuses through the cell membrane and binds to and subsequently activates the nuclear estrogen receptor found in the reproductive tract, breast, pituitary, hypothalamus, liver, and bone. The activated hormone-receptor complex gets translocated to the nucleus where it binds to the estrogen response element on the dna and promotes the transcription of target genes involved in the functioning of the female reproductive system and secondary sex characteristics. Estrone increases the hepatic synthesis of sex hormone binding globulin (shbg), thyroid-binding globulin (tbg), and other serum proteins, as well as suppresses the release of follicle-stimulating hormone (fsh) and luteinizing hormone (lh) from the anterior pituitary. [National Center for Biotechnology Information. PubChem Compound Database; CID=5870].
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 31.25 to 2000 pg/mL
Assay Sensitivity 22.4 pg/mL
Description Estrone Competitive ELISA Kit
Assay Multiplex assay on the Luminex Platform
Conjugate HRP
Detection Method Fluorescence
Product Type ELISA
Protein Family Steroid hormone
Sample Type Fecal Extract, Supernatant, Urine
Sufficient For 96 assays
Target Species Human
For Use With (Application) ELISA, Protein Assays, Aktivity Assays, Protein Biology, Reproductive Biology, Inflammation, Oxidative Stress, Metabolism, Cell Signaling, Epigenetics, Hematology, DNA Damage, Cancer Biology, Immunology, Neuroscience, Neurobiology, Steroid Hormone Detecti
For Use With (Equipment) Colorimetric Microplate Reader
Interassay CV 5.5%
Intraassay CV 4.7
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, HRP-Conjugated Detection Antibody, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 96 Tests
Regulatory Status RUO
Research Discipline Reproductive Biology
Sample Volume Fecal Extract 50 μL, Supernatant 50 μL, Urine 12.5 μL
Storage Requirements -20°C
Target Estrone
Test Time 1 hrs 20 mins
Total Assay Time 2 hrs 30 mins
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.