Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ESYT2/FAM62B Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501981019
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-1019 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-1019 Supplier Bioss Supplier No. BS11003R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Tethers the endoplasmic reticulum to the cell membrane and promotes the formation of appositions between the endoplasmic reticulum and the cell membrane. Binds glycerophospholipids in a barrel-like domain and may play a role in cellular lipid transport. Plays a role in FGF signaling via its role in the rapid internalization of FGFR1 that has been activated by FGF1 binding; this occurs most likely via the AP-2 complex.

Specifications

Antigen ESYT2/FAM62B
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene ESYT2
Gene Accession No. A0FGR8
Gene Alias 2410017M09Rik; 4921504I16Rik; chr2 synaptotagmin; Chr2Syt; D12Ertd551e; ESYT2; E-Syt2; extended synaptotagmin 2; extended synaptotagmin like protein 2; extended synaptotagmin protein 2; Extended synaptotagmin-2; extended synaptotagmin-like protein 2; FAM62B; family with sequence similarity 62 (C2 domain containing), member B; family with sequence similarity 62, member B; KIAA1228; RGD1565705
Gene Symbols ESYT2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human ESYT2/FAM62B.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57488
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.