Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ETAA1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557129
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557129 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557129 Supplier Invitrogen™ Supplier No. PA557129
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: INNNSEHGAK NMFAISKQGS NLVQSKHLNP GSISVQTSLT NSSQIDKPMK MEKGEMYGNS PRFLGATNLT MYSKISNCQI NNLHVSYTNT DVPIQVN Highest antigen sequence identity to the following orthologs: Mouse - 36%, Rat - 44%.

ETAA1 (Ewing's tumor-associated antigen 1), also known as ETAA16, is a 926 amino acid cytoplasmic protein that is highly expressed in kidney, brain, liver and Ewing tumor cell lines. ETAA1 undergoes post-translational phosphorylation following DNA damage, most likely by either ATM or ATR, and is suggested to function as a tumour-specific cell surface antigen in Ewing's family of tumour cell lines. The gene encoding ETAA1 maps to human chromosome 2, which consists of 237 million bases, encodes over 1,400 genes and makes up approximately 8% of the human genome. A number of genetic diseases are linked to genes on chromosome 2 including Harlequin icthyosis, sitosterolemia and Alstrom syndrome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ETAA1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ETAA1
Gene Accession No. Q9NY74
Gene Alias 5730466H23Rik; AA672646; Etaa1; ETAA16; Ewing tumor associated antigen 1; Ewing tumor-associated antigen 1; ewing's tumor-associated antigen 1; Ewing's tumor-associated antigen 1 homolog; ewing's tumor-associated antigen 16; RGD1560957
Gene Symbols ETAA1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ETAA1. Recombinant protein control fragment (Product #RP-102644).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54465
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.