Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Etanercept Mouse anti-Human, Clone: ETA63C8, MilliporeSigma™

Catalog No. MABF197325 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF1973-25 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABF1973-25 Supplier MilliporeSigma Supplier No. MABF197325UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This mouse monoclonal Anti-Etanercept, clone ETA63C8 Antibody for the detection of Etanercept.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Etanercept
Applications Neutralization, ELISA
Classification Monoclonal
Clone ETA63C8
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Host Species Mouse
Immunogen Chimeric fusion protein cotaining the extracellular ligand-binding portion of the p75 TNF receptor linked to the Fc portion of human IgG1.
Purification Method Protein G Purified
Quantity 25 μL
Regulatory Status RUO
Research Discipline Inflammation and Immunology
Primary or Secondary Primary
Test Specificity Clone ETA63C8 detects Etanercept in human samples. It can bind to Etanercept and block its activity.
Target Species Human
Content And Storage Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.