Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ETEA Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5111672
Change view
Click to view available options
Quantity:
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5111672 100 μL
PIPA5111715 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PIPA5111672 Supplier Invitrogen™ Supplier No. PA5111672
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EGYRVSQALR ENTYPFLAMI MLKDRRMTVV GRLEGLIQPD DLINQLTFIM DANQTYLVSE RLEREERNQT QVL.

The protein encoded by this gene is highly expressed in peripheral blood of patients with atopic dermatitis (AD), compared to normal individuals. It may play a role in regulating the resistance to apoptosis that is observed in T cells and eosinophils of AD patients.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ETEA
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FAF2
Gene Accession No. Q96CS3
Gene Alias 2210404d11rik; AI462440; ETEA; expressed in T-cells and eosinophils in atopic dermatitis; FAF2; Fas associated factor family member 2; FAS-associated factor 2; Kiaa0887; mKIAA0887; Protein ETEA; protein expressed in T-cells and eosinophils in atopic dermatitis; RGD1306577; UBX domain containing 8; UBX domain protein 3B; UBX domain-containing protein 3B; UBX domain-containing protein 8; UBX8; UBXD8; UBXN3B; Unknown (protein for MGC:129072)
Gene Symbols FAF2
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human ETEA. Recombinant protein control fragment (Product #RP-108582).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23197
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.