Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ETF1 Mouse anti-Human, Clone:4F9H12, Abnova™
Mouse monoclonal antibody raised against recombinant human ETF1.
Supplier: Abnova Corporation MAB17190
Description
Termination of protein biosynthesis and release of the nascent polypeptide chain are signaled by the presence of an in-frame stop codon at the aminoacyl site of the ribosome. The process of translation termination is universal and is mediated by protein release factors (RFs) and GTP. A class 1 RF recognizes the stop codon and promotes the hydrolysis of the ester bond linking the polypeptide chain with the peptidyl site tRNA, a reaction catalyzed at the peptidyl transferase center of the ribosome. Class 2 RFs, which are not codon specific and do not recognize codons, stimulate class 1 RF activity and confer GTP dependency upon the process. In prokaryotes, both class 1 RFs, RF1 and RF2, recognize UAA; however, UAG and UGA are decoded specifically by RF1 and RF2, respectively. In eukaryotes, eRF1, or ETF1, the functional counterpart of RF1 and RF2, functions as an omnipotent RF, decoding all 3 stop codons (Frolova et al., 1994 [PubMed 7990965]).[supplied by OMIM]Specifications
ETF1 | |
Monoclonal | |
Unconjugated | |
ELISA (1:10000) Western Blot (1:500-1:2000) Immunohistochemistry Immunocytochemistry Flow Cytometry (1:200-1:400) The optimal working dilution should be determined by the end user. |
|
ETF1 | |
Mouse | |
100 μg | |
Human | |
Antibody | |
IgG1 |
ELISA, Flow Cytometry, Western Blot | |
4F9H12 | |
eukaryotic translation termination factor 1 | |
In PBS (0.05% sodium azide) | |
D5S1995, ERF, ERF1, MGC111066, RF1, SUP45L1, TB3-1 | |
Recombinant protein corresponding to amino acid 288-437 of human ETF1 from E. coli. | |
2107 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction