Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ETFA Antibody (OTI5C4), Alexa Fluor™ 750, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270693AF750
Description
ETFA Monoclonal specifically detects ETFA in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
ETFA | |
Monoclonal | |
Alexa Fluor 750 | |
50 mM sodium borate with 0.05% sodium azide | |
ETFA | |
Full length recombinant protein of human ETFA (NP_000117) produced in E.coli. | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Western Blot | |
OTI5C4 | |
Western Blot | |
alpha-ETF, electron transfer flavoprotein alpha-subunit, Electron transfer flavoprotein subunit alpha, mitochondrial, electron-transfer-flavoprotein, alpha polypeptide, EMA, GA2, glutaric aciduria II, MADD | |
Mouse | |
0.1 mL | |
2108 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction