Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ETFB Polyclonal Antibody

Goat Polyclonal Antibody
Supplier: Invitrogen™ PA519336
Description
This antibody is predicted to react with bovine and canine based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 8,000.
This gene encodes electron-transfer-flavoprotein, beta polypeptide, which shuttles electrons between primary flavoprotein dehydrogenases involved in mitochondrial fatty acid and amino acid catabolism and the membrane-bound electron transfer flavoprotein ubiquinone oxidoreductase. The gene deficiencies have been implicated in type II glutaricaciduria. Alternatively spliced transcript variants have been found for this gene.
Specifications
| ETFB | |
| Polyclonal | |
| Unconjugated | |
| ETFB | |
| 0610009I16Rik; 2810441H06Rik; beta-ETF; electron transfer flavoprotein beta subunit; electron transfer flavoprotein beta-subunit; electron transfer flavoprotein subunit beta; electron transfer flavoprotein, beta polypeptide; electron transferring flavoprotein, beta polypeptide; electron-transfer-flavoprotein, beta polypeptide; electron-transferring-flavoprotein, beta polypeptide; Etfb; FP585; MADD | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 110826, 2109, 292845 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| P38117, Q68FU3, Q9DCW4 | |
| ETFB | |
| Synthetic peptide sequence (QVTLEGDKLKVERE) corresponding to the internal amino acids of ETFB (aa 152-165). | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction