Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ETFB Polyclonal Antibody, Biotin
GREENER_CHOICE

Catalog No. PIPA5142591
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5142591 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5142591 Supplier Invitrogen™ Supplier No. PA5142591
Add to Cart
Add to Cart

Goat Polyclonal Antibody

Additional Information: This antibody is expected to recognize both reported isoforms (NP_001976.1; NP_001014763.1). This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:8,000.

This gene encodes electron-transfer-flavoprotein, beta polypeptide, which shuttles electrons between primary flavoprotein dehydrogenases involved in mitochondrial fatty acid and amino acid catabolism and the membrane-bound electron transfer flavoprotein ubiquinone oxidoreductase. The gene deficiencies have been implicated in type II glutaricaciduria. Alternatively spliced transcript variants have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ETFB
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Biotin
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene ETFB
Gene Accession No. P38117, Q68FU3, Q9DCW4
Gene Alias 0610009I16Rik; 2810441H06Rik; beta-ETF; electron transfer flavoprotein beta subunit; electron transfer flavoprotein beta-subunit; electron transfer flavoprotein subunit beta; electron transfer flavoprotein, beta polypeptide; electron transferring flavoprotein, beta polypeptide; electron-transfer-flavoprotein, beta polypeptide; electron-transferring-flavoprotein, beta polypeptide; Etfb; FP585; MADD
Gene Symbols ETFB
Host Species Goat
Immunogen Peptide with sequence C-PQRTAGVKVETTE (aa 231-243).
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 110826, 2109, 292845
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.