Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ETFDH Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA519340
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA519340 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA519340 Supplier Invitrogen™ Supplier No. PA519340
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is predicted to react with bovine, canine and rabbit based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 2,000.

Electron-transferring-flavoprotein dehydrogenase in the inner mitochondrial membrane accepts electrons from electron-transfer flavoprotein which is located in the mitochondrial matrix and reduces ubiquinone in the mitochondrial membrane. The protein is synthesized as a 67-kDa precursor which is targeted to mitochondria and processed in a single step to a 64-kDa mature form located in the mitochondrial membrane. Deficiency in electron-transferring-flavoprotein dehydrogenase have been demonstrated in some patients with type II glutaricacidemia.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ETFDH
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene Etfdh
Gene Accession No. Q16134, Q6UPE1, Q921G7
Gene Alias 0610010I20Rik; AV001013; electron transfer flavoprotein dehydrogenase; electron transfer flavoprotein-ubiquinone oxidoreductase, mitochondrial; electron transferring flavoprotein dehydrogenase; electron transferring flavoprotein, dehydrogenase; electron-transferring-flavoprotein dehydrogenase; ETF dehydrogenase; Etfdh; ETFQO; ETF-QO; ETF-ubiquinone oxidoreductase; MADD
Gene Symbols Etfdh
Host Species Goat
Immunogen Synthetic peptide sequence (EHDQPAHLTLRD) corresponding to the internal amino acids of ETFDH (aa 530-541).
Purification Method Ammonium Sulfate Precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2110, 295143, 66841
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.