Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ETFDH Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559808
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559808 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559808 Supplier Invitrogen™ Supplier No. PA559808
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LAVAHEKDIR VCLVEKAAQI GAHTLSGACL DPGAFKELFP DWKEKGAPLN TPVTEDRFGI LTEKYRIPVP ILPGLPMN Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 92%.

Electron-transferring-flavoprotein dehydrogenase in the inner mitochondrial membrane accepts electrons from electron-transfer flavoprotein which is located in the mitochondrial matrix and reduces ubiquinone in the mitochondrial membrane. The protein is synthesized as a 67-kDa precursor which is targeted to mitochondria and processed in a single step to a 64-kDa mature form located in the mitochondrial membrane. Deficiency in electron-transferring-flavoprotein dehydrogenase have been demonstrated in some patients with type II glutaricacidemia.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ETFDH
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Etfdh
Gene Accession No. Q16134
Gene Alias 0610010I20Rik; AV001013; electron transfer flavoprotein dehydrogenase; electron transfer flavoprotein-ubiquinone oxidoreductase, mitochondrial; electron transferring flavoprotein dehydrogenase; electron transferring flavoprotein, dehydrogenase; electron-transferring-flavoprotein dehydrogenase; ETF dehydrogenase; Etfdh; ETFQO; ETF-QO; ETF-ubiquinone oxidoreductase; MADD
Gene Symbols Etfdh
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ETFDH. Recombinant protein control fragment (Product #RP-96739).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2110
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.