Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ETFDH Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA598423
Description
ETFDH Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA
Electron-transferring-flavoprotein dehydrogenase in the inner mitochondrial membrane accepts electrons from electron-transfer flavoprotein which is located in the mitochondrial matrix and reduces ubiquinone in the mitochondrial membrane. The protein is synthesized as a 67-kDa precursor which is targeted to mitochondria and processed in a single step to a 64-kDa mature form located in the mitochondrial membrane. Deficiency in electron-transferring-flavoprotein dehydrogenase have been demonstrated in some patients with type II glutaricacidemia.
Specifications
ETFDH | |
Polyclonal | |
Unconjugated | |
Etfdh | |
0610010I20Rik; AV001013; electron transfer flavoprotein dehydrogenase; electron transfer flavoprotein-ubiquinone oxidoreductase, mitochondrial; electron transferring flavoprotein dehydrogenase; electron transferring flavoprotein, dehydrogenase; electron-transferring-flavoprotein dehydrogenase; ETF dehydrogenase; Etfdh; ETFQO; ETF-QO; ETF-ubiquinone oxidoreductase; MADD | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
2110 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q16134 | |
Etfdh | |
Recombinant Human Electron transfer flavoprotein-ubiquinone oxidoreductase, mitochondrial protein (318-617AA). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction