Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethanesulfonyl chloride, 98+%

Catalog No. AC215820025
Click to view available options
Quantity:
5 mL
50 mL
100 mL
500 mL
2.5 L
Packaging:
Glass bottle

CAS: 594-44-5 | C2H5ClO2S | 128.57 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 594-44-5
Molecular Formula C2H5ClO2S
Molecular Weight (g/mol) 128.57
MDL Number MFCD00007460
InChI Key FRYHCSODNHYDPU-UHFFFAOYSA-N
Synonym ethylsulfonyl chloride, ethylsulfochloride, 1-ethanesulfonyl chloride, ethanesulphonyl chloride, ethanesulfonylchloride, unii-b56zoc898a, ethylsulfonylchloride, ethansulfonyl chloride, ethansulfonylchlorid, ethanesulfonylchoride
PubChem CID 11667
IUPAC Name ethanesulfonyl chloride
SMILES CCS(Cl)(=O)=O

Specifications

Color Brown to Pink-Purple or Colorless to Yellow
Density 1.3500g/mL
Boiling Point 177.0°C
Flash Point 83°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Refractive Index 1.4520 to 1.4540
Linear Formula C2H5SO2Cl
Quantity 2.5 L
Beilstein 04, 6
Specific Gravity 1.35
Solubility Information Solubility in water: decomposes in water. Other solubilities: soluble in acetone and ethyl ether
Viscosity 1.44 mm2/s (20°C)
Formula Weight 128.58
Percent Purity 98+%
Physical Form Liquid
Chemical Name or Material Ethanesulfonyl chloride, 98+%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Toxic if inhaled.
Harmful if swallowed.
Causes serious eye damage.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 209-842-5
RTECSNumber KI8050000
TSCA TSCA
Recommended Storage this product darkens during storage

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.