Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethanethiol, 97%

Catalog No. AA22585AE
Click to view available options
Quantity:
25 mL
100 mL
500 mL

CAS: 75-08-1 | C2H6S | 62.13 g/mol

Ethanethiol acts as a stabilizer for adhesives and an odorant for natural gas. It is used in the odorless liquefied petroleum gas (LPG) in order to find out the gas leaks. It is used as key starting material for the preparation of ethylsulfonic acid, diethyldisulfide and sodium ethylmercaptide. Further, it is also used in the underground mining industry as stench gas. In organic synthesis, it gives powerful nucleophile set- in the presence of sodium hydroxide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-08-1
Molecular Formula C2H6S
Molecular Weight (g/mol) 62.13
MDL Number MFCD00004887
InChI Key DNJIEGIFACGWOD-UHFFFAOYSA-N
Synonym ethyl mercaptan, mercaptoethane, thioethanol, ethyl hydrosulfide, ethyl sulfhydrate, ethyl thioalcohol, thioethyl alcohol, 1-mercaptoethane, aethanethiol, aethylmercaptan
PubChem CID 6343
IUPAC Name ethanethiol
SMILES CCS

Specifications

Melting Point -148°C
Color Colorless
Density 0.84
Boiling Point 35°C
Flash Point −48°C (−55°F)
Odor Unpleasant
Refractive Index 1.431
Linear Formula CH3CH2SH
Quantity 100 mL
UN Number UN2363
Beilstein 773638
Merck Index 14,3726
Solubility Information Soluble in alcohol,ether,waterMiscible with alcohol,petroleum naphtha,acetone,dilute alkali and ether; Slightly miscible with water.
Formula Weight 62.13
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Ethanethiol
Show More Show Less
Hazard Category H225-H302+H332-H317
Hazard Statement GHS H Statement
H225-H332;
Highly flammable liquid and vapor.
Harmful if inhaled.
Precautionary Statement P210-P233-P240-P241-P242-P243-P261-P264b-P270-P271-P272-P280-P301+P312-P303+P361+P353-P304+P340-P312-P330-P333+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: I; Proper Shipping Name: ETHYL MERCAPTAN
EINECSNumber 200-837-3
RTECSNumber KI9625000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.