Alfa Aesar™ Ethanethiol, 97%

Manufacturer: Alfa Aesar™ 22585AC

Quantity 25mL

 View more versions of this product

Catalog No. AA22585AC

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 75-08-1
    Linear Formula CH3CH2SH
    Formula Weight 62.13
    MDL Number MFCD00004887
    UN Number UN2363
    Beilstein 773638
    Merck Index 14,3726
    Synonym Ethyl mercaptan
    Chemical Name or Material Ethanethiol

    Specifications

    Formula Weight 62.13
    Health Hazard 3 GHS P Statement
    P210-P261-P280-P303+P361+P353-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    Store in a well-ventilated place.
    Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Odor Unpleasant
    Linear Formula CH3CH2SH
    Synonym Ethyl mercaptan
    Density 0.84g/mL
    Color Colorless
    TSCA TSCA
    MDL Number MFCD00004887
    UN Number UN2363
    Merck Index 14,3726
    Solubility Information Soluble in alcohol, ether, water
    Health Hazard 1 Danger
    Refractive Index 1.4310
    Boiling Point 35°C
    Flash Point −48° (−55°F)
    DOT Information Transport Hazard Class: 3; Packing Group: I; Proper Shipping Name: ETHYL MERCAPTAN
    Physical Form Liquid
    Beilstein 773638
    Melting Point −148°C
    Quantity 25mL
    CAS 75-08-1
    Health Hazard 2 GHS H Statement
    H225-H332
    Highly flammable liquid and vapor.
    Harmful if inhaled.
    RTECS Number KI9625000
    Chemical Name or Material Ethanethiol
    Assay 97%

    GHS H Statement
    H225-H332
    Highly flammable liquid and vapor.
    Harmful if inhaled.

    GHS P Statement
    P210-P261-P280-P303+P361+P353-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    Store in a well-ventilated place.
    Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit