Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethanol, 190 proof (95%), Tax-Paid, Decon™ Labs
GREENER_CHOICE

Catalog No. 04355461
Click to view available options
Quantity:
1 gal.
5 gal.
4 x 1 gal.
55 gal.
Packaging:
Drum
Glass Bottle
Metal Container
Plastic Bottle
Poly Bottle with Spigot Closure

Federal Tax is included in the price. No federal permit required for purchase

Used for research, healthcare, industrial, or production applications as a cleaner, solvent or reagent.

Chemical Identifiers

CAS 64-17-5, 7732-18-5
Molecular Formula C2H6O
Molecular Weight (g/mol) 46.07 g/mol
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
Synonym ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol
PubChem CID 702
ChEBI CHEBI:16236
IUPAC Name Ethanol
SMILES CCO

Specifications

Melting Point -114°C
CAS Min % 94.9%
Color Clear
CAS Max % 96%
Density 6.72
Boiling Point 78.3°C
Quantity 55 gal.
Packaging Metal Container
Physical Form Liquid
UN Number UN1170
Percent Purity 95%
Grade USP
Chemical Name or Material Ethanol, 190 Proof (95%)
Show More Show Less
DOTInformation DOT Class: 3, Packaging: II
Recommended Storage Do not store alcohol above 120°F
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.