Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ethanol, 200 proof (100%), Tax-Paid, Decon™ Labs
Shop All Decon Labs Products

Click to view available options
Quantity:
0.125 gal.
1 gal.
5 gal.
4 x 1 gal.
55 gal.
24 x 0.125 gal.
Packaging:
Drum
Glass Bottle
Metal Container
Plastic Bottle
Poly Bottle
Description
Used for research, healthcare, industrial, or production applications as a cleaner, solvent or reagent.
Specifications
Specifications
Melting Point | -114°C |
CAS Min % | 99.9% |
Color | Clear |
CAS Max % | 100% |
Density | 6.62 |
Boiling Point | 78.3°C |
Quantity | 55 gal. |
Packaging | Metal Container |
Physical Form | Liquid |
UN Number | UN1170 |
Show More |
Safety and Handling
DOTInformation | DOT Class: 3, Packaging II |
Recommended Storage | Do not store alcohol above 120°F. |
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction