Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethanol, 70% Solution, Molecular Biology Grade, Denatured, Fisher BioReagents™ PROMO

Catalog No. BP82031GF25 Shop All Fisherbrand Products
Click to view available options
:
1 gal.

Used as a cleaning agent for laboratory use.

  • DNase, and Protease tested to verify absence of hydrolytic enzymes
  • Combine this blend with commercial 'mini-prep' kits or traditional extraction
  • ACS grade Absolute Ethanol used in preparation of solution
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64-17-5, 67-63-0, 7732-18-5
Molecular Formula C2H6O
Molecular Weight (g/mol) 46.069 g/mol
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
Synonym ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol
PubChem CID 702
ChEBI CHEBI:16236
IUPAC Name ethanol
SMILES CCO

Specifications

Melting Point -144°C
Color Colorless
Boiling Point 78.4°C
Quantity 1 gal.
Packaging Poly Bottle
Physical Form Liquid
Residue after Evaporation ≤ 0.001%
Vapor Pressure 44.6 mm Hg at 20°C (68°F)
Water 3% to 7% (GC-TCD)
Grade Molecular Biology
Chemical Name or Material 70% Denatured Ethanol Solution (Molecular Biology Grade)
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.