Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethanol, 95% Denatured, Vion Biosciences
SCI_ED

Catalog No. S02505 Shop All Aldon Products
Change view
Click to view available options
Quantity:
1 L
3.8 L
4 x 1 gal.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S02505 4 x 1 gal.
S02507 1 L
S02506 3.8 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
WARNING: This product can expose you to chemicals including Methanol and Methyl isobutyl ketone, which are known to the State of California to cause cancer and birth defects or other reproductive harm.
Catalog No. S02505 Supplier Aldon Corporation Supplier No. VNEE0080CS/4
Add to Cart
Add to Cart
WARNING: This product can expose you to chemicals including Methanol and Methyl isobutyl ketone, which are known to the State of California to cause cancer and birth defects or other reproductive harm.

This 95% denatured ethanol is a high-quality fixative and dehydrant that allows for optimal staining in histology applications.

  • This 95% denatured ethanol is a high-quality fixative and dehydrant that allows for optimal staining in histology applications.

Chemical Identifiers

CAS 67-63-0 , 7732-18-5 , 64-17-5 , 67-56-1 , 108-10-1
Molecular Formula C3H8O
Molecular Weight (g/mol) 60.10
MDL Number MFCD00003568
InChI Key KFZMGEQAYNKOFK-UHFFFAOYSA-N
Synonym Ethanol, Denatured, 95%
PubChem CID 702
ChEBI CHEBI:15235
IUPAC Name propan-2-ol
SMILES CC(C)O

Specifications

pH Data not available.
Melting Point -114°C
CAS Min % 8.55%, 0.85%, 80.75%, 3.80%, 5.00%
Color Colorless
CAS Max % 81.51%, 4.08%, 0.95%, 5.00%, 8.55%
Appearance Clear, colorless liquid.
Density 0.7919 to 0.7955 g/mL
Quantity 4 x 1 gal.
Explosion Limit 4.00%
Flash Point 5°C
Identification 95% Ethanol
Ignition Point 400°C
Infrared Spectrum Yes
Odor Mild Characteristic Odor
Packaging 1 Gallon F Style UN Bottle
Physical Form Liquid
Product Line Vion Histology
Vapor Pressure Ca 50 (at 20°C)
Linear Formula C2H5OH
UN Number UN1170
Water 5.00%
Concentration 95%
Formulation 95% Denatured Ethanol
Boiling Point 74°C to 80°C
Solubility Information Soluble in water.
InChI Formula 1S/C2H6O/c1-2-3/h3H,2H2,1H3
Percent Purity 80.75 to 81.51%
Grade Histological
Chemical Name or Material Ethanol
Show More Show Less
Safety handling Check Alert storage code Red
Hazard Category Toxic if swallowed.
Hazard Statement Toxic if in contact with skin.
Precautionary Statement Causes serious eye irritation.
ShelfLife 3 Years
DOTInformation UN1170, Ethanol, 3(II), Limited quantity equal to or less than 1 L
EINECSNumber 200-578-6
RTECSNumber KQ6300000
Recommended Storage Store in a cool, dry, well-ventilated area away from incompatible substances. Keep away from ignition sources.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.