Learn More
Ethanol, 99.5%, ACS reagent, absolute, 200 proof

Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications
Specifications
Density | 0.7900g/mL |
Quantity | 2 L |
Infrared Spectrum | Conforms to reference |
Packaging | N/A |
Refractive Index | 1.3585 to 1.3605 |
Assay Percent Range | 99.5% |
Residue after Evaporation | 0.001% max. |
Titratable Acid | 0.0005 meq/g max. |
Titratable Base | 0.0002 meq/g max. |
Linear Formula | CH3CH2OH |
Show More |
Safety and Handling
Hazard Category | Danger |
Hazard Statement | GHS H Statement: Highly flammable liquid and vapor. |
Precautionary Statement | GHS P Statement: Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Ground/Bond container and receiving equipment. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification. |
For Manufacturing and Laboratory Use. Not for beverage use
Your input is important to us. Please complete this form to provide feedback related to the content on this product.