Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethanol, Absolute (200 Proof), Molecular Biology Grade, Fisher BioReagents™
GSA_VA

Catalog No. p-3759149 Shop All Fisherbrand Products
Click to view available options
Quantity:
100 mL
500 mL
4 L

Meets ACS Specifications

  • Meets ACS Specifications
  • For nucleic acid purification and precipitation
  • Ultrapure molecular biology grade ethanol is used for the purification and precipitation of biomolecules such as nucleic acids and proteins
  • It is also used in histology to prepare staining and destaining reagents and for dehydrating tissues prior to embedding

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64-17-5
Molecular Formula C2H6O
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
Synonym ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol
PubChem CID 702
ChEBI CHEBI:16236
IUPAC Name ethanol
SMILES CCO

Specifications

Melting Point -114°C
Color Colorless
Boiling Point 78°C
Quantity 100 mL
Flash Point 12°C (53.6°F)
Identification Pass Test
Infrared Spectrum Conforms to reference
Physical Form Liquid
Residue after Evaporation 0.001% max.
Assay Percent Range Assay: ≥99.5% (v/v)
Titratable Acid 0.0005 mEq /g max.
Titratable Base 0.0002 mEq /g max.
Linear Formula C2H5OH
Water 0.2% max.
Merck Index 15, 3814
Molecular Weight (g/mol) 46.069
Formula Weight 46.07
Percent Purity ≥99.5%
Grade DNase-, RNase- and Protease-Free
Chemical Name or Material Ethanol, Absolute (200 Proof)
Show More Show Less
Hazard Statement WARNING!
Precautionary Statement Emergency Overview
Flammable liquid and vapor. May cause irritation of respiratory tract. May cause central nervous system effects. Aspiration hazard if swallowed - can enter lungs and cause damage. This substance has caused adverse reproductive and fetal effects in humans. Substances known to cause developmental toxicity in humans. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Obtain medical attention. Move to fresh air.

NFPA

Health:2
Flammability:3
Instability:0
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.