Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethanol, Alcohol Reagent, anhydrous, denatured, ACS, 94-96%

Catalog No. p-4408250
Click to view available options
Quantity:
500 mL
1 L
4 L
4 x 1 L

CAS: 64-17-5 | C2H6O | 46.069 g/mol

Ethanol is used in antibacterial hand sanitizer gels, as an antidote to methanol poisoning and an antitussive agent. It is acting as a solvent to remove the paint and ink stain on curtains, dress and upholstery. It is utilized as a fuel in camping and primus stoves. It is likewise employed to clean the windows, manufacturing nail-polish and to test pectin levels in jam and jelly making. It finds application in personal care products such as mouthwashes, perfumes and deodorants.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64-17-5
Molecular Formula C2H6O
Molecular Weight (g/mol) 46.069
MDL Number MFCD00003568
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
Synonym ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol
PubChem CID 702
ChEBI CHEBI:16236
IUPAC Name ethanol
SMILES CCO

Specifications

Melting Point −90°C
Density 0.789 g/mL
Boiling Point 78°C
Quantity 1 L
Flash Point 11°C (52°F)
Physical Form Liquid
Refractive Index 1.361
Assay Percent Range 94-96%
Beilstein 1718733
Solubility Information Miscible with water,methanol,ether,chloroform and acetone.
Formula Weight 46.08
Grade ACS Reagent
Chemical Name or Material Ethanol, Alcohol Reagent, anhydrous, denatured, For ACS analysis; Begins to lose water at 60°, anhydrous at 100°
Show More Show Less
Hazard Category H225-H302+H332-H319-H336-H371
Precautionary Statement P210-P233-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P309+P311-P312-P330-P370+P378q-P501c
DOTInformation Hazard Class: 3; Packaging Group: II
EINECSNumber 200-578-6
RTECSNumber KQ6300000
TSCA Yes
Recommended Storage Ambient temperatures

For Manufacturing and Laboratory Use. Not for beverage use

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.