Learn More
Ethanol, anhydrous, denatured, 85.8%, 13.3% methanol, 0.9% MIBK
Description
Denatured ethanol is acting as a solvent to remove the paint and ink stain on curtains, dress and upholstery. It is utilized as fuel in camping and primus stoves. It is likewise employed to clean the windows, manufacturing nail-polish and to test pectin levels in jam and jelly making.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- As a gasoline additive/substitute; octane booster
- Very mobile liquid solvent
- Used in lotions, tonics and perfumes
Specifications
Specifications
| Melting Point | -100°C |
| Color | Colorless |
| Density | 0.811 g/mL |
| Boiling Point | 80°C |
| Quantity | 20 L |
| Flash Point | 11°C (52°F) |
| Odor | Like alcohol |
| Physical Form | Liquid |
| Refractive Index | 1.361 |
| Assay Percent Range | 85.8% |
| Show More |
Safety and Handling
| Hazard Category | H225-H302-H319-H331-H370 |
| Hazard Statement | GHS H Statement H225-H370 Highly flammable liquid and vapor. Causes damage to organs. |
| Precautionary Statement | P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P307+P311-P311-P330-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S. |
| EINECSNumber | 200-578-6 |
| RTECSNumber | KQ6300000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
For Manufacturing and Laboratory Use. Not for beverage use
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.