Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethanol-d{6}, anhydrous, 99+% (Isotopic)

Catalog No. p-7049638
Click to view available options
Quantity:
1 g
5 g

NMR Solvent | CAS: 1516-08-1 | C2H6O | 52.106 g/mol

Ethanol-d_6, is employed as an NMR solvent. It is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1516-08-1
Molecular Formula C2H6O
Molecular Weight (g/mol) 52.106
MDL Number MFCD00051020
InChI Key LFQSCWFLJHTTHZ-LIDOUZCJSA-N
Synonym ethanol-d6, 2h6 ethanol, hexadeuteroethanol, 2 h? ethan 2 h ol, ethyl-d5 alcohol-d, ethyl alcohol-d6, ethanol d, ethanol-d6 9ci
PubChem CID 102138
IUPAC Name 1,1,1,2,2-pentadeuterio-2-deuteriooxyethane
SMILES CCO

Specifications

Physical Form Liquid
Color Colorless
Quantity 1 g
Density 0.91 g/mL
Sensitivity Hygroscopic
Odor Alcohol-like
Boiling Point 79°C
Linear Formula CD3CD2OD
UN Number UN1170
Solubility Information Soluble in water.
Formula Weight 52.11
Percent Purity ≥99%
Chemical Name or Material Ethanol-d6, Isotopic, Anhydrous
Show More Show Less
Hazard Category H224
Hazard Statement GHS H Statement
H225
Highly flammable liquid and vapor.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ETHANOL SOLUTIONS
EINECSNumber 216-162-2
TSCA No
Recommended Storage Ambient

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.