Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethanol, denatured, special, Thermo Scientific™

Catalog No. AA41603K2
Change view
Click to view available options
Quantity:
1 L
4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA41603K2 1 L
AA41603K7 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA41603K2 Supplier Thermo Scientific Chemicals Supplier No. 41603K2

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64-17-5 , 67-56-1 , 141-78-6
Molecular Formula C2H6O
Molecular Weight (g/mol) 46.069
MDL Number MFCD00003568
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
Synonym ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol
PubChem CID 702
ChEBI CHEBI:16236
IUPAC Name ethanol
SMILES CCO

Specifications

Color Colorless
Appearance Colorless liquid
Density 0.7893g/mL
Quantity 1 L
Flash Point 13°C (55°F)
Odor Alcohol-like
Physical Form Liquid
Refractive Index 1.361
Beilstein 1718733
Solubility Information Miscible in water.
Formula Weight 46.08
Concentration or Composition (by Analyte or Components) Ethyl alcohol: 92%; Methanol: 4%; Ethyl acetate: 4%
Chemical Name or Material Ethanol
Show More Show Less
Hazard Category H225-H319-H336-H371
Precautionary Statement P210-P233-P240-P241-P242-P243-P260-P264b-P270-P280-P303+P361+P353-P305+P351+P338-P309+P311-P370+P378q-P501c
DOTInformation DOT Class: 3, Packaging Group: II
EINECSNumber 200-578-6
RTECSNumber KQ6300000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.