Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Ethanol (EtOH), 20%

Catalog No. p-200003983
Click to view available options
Packaging:
Drum
Sample
Tote
Quantity:
5 L
20 L
200 L

Gibco 20% Ethanol (EtOH) is sterile-filtered, packaged in bioprocess containers, and stocked to be available when you need it. Sourcing 20% EtOH can streamline your workload for equipment sanitization and column storage, allowing you to focus on the key steps in your downstream processes.

Ethanol (EtOH) and bulk Water for Injection (WFI) are mixed to produce solutions that meet the most stringent quality control expectations in accordance with the pharmaceutical, biopharma, and diagnostic industries. Gibco 20% Ethanol is manufactured to cGMP standards at an ISO certified facility, promoting quality and convenience for every stage of a biologic’s pipeline.

Product features:
• Available in a range of pack sizes (5 L, 20 L, 200 L)
• Product specifications are well-defined, with the established solutions ready to order
• Manufactured to ISO and cGMP standards to meet the stringent expectations of highly regulated industries
• Readily accessible documentation for accelerated qualification

User benefits:
• Meets workflow needs for all stages of biologic development and manufacturing
• Streamlines in-house processes related to formulation, hydration, testing, and validation
• Mitigates space required for buffer preparation and storage
• Eases the time requirements dedicated for in-house buffer preparation

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ethyl Alcohol
Color Clear
Quantity 5 L
Packaging Sample
Physical Form Liquid
Certifications/Compliance GMP - 21 CFR 820, ISO 13485
Concentration 20%
Sterilization Method 0.2 μm Sterile-filtered
Formulation 20% EtOH

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.