Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethanol Fuel Cell Science Kit
SCI_ED

Catalog No. s12822
Change view
Click to view available options
:
Ethanol fuel cell science kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
S12822 Ethanol fuel cell science kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S12822 Supplier Horizon Fuel Cell Technologies Supplier No. FCJJ42
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Includes

  • Ethanol/water mix container
  • Fuel tank with lid
  • Ethanol fuel cell
  • Electric fan loads
  • Silicon tubes
  • pH paper
  • Clamp
  • Renewable Energy Curriculum CD

Miniature power plant where ethanol is the feedstock.

Fuel cell uses hydrogen in the ethanol to produce electricity without combustion.

  • Understanding the workings of an ethanol fuel cell
  • pH paper shows change of property of the consumed ethanol fuel
  • Fuel solution can be made with household items
  • Weight: 0.50kg (1.1 lb.)

Specifications

For Use With (Application) Demonstrate the workings of an ethanol fuel cell
Includes Ethanol/water mix container; Fuel tank with lid; Ethanol fuel cell; Electric fan loads; Silicon tubes; pH paper; Clamp; Renewable Energy Curriculum CD
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.